The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271353.1 | NP_001258282.1 | 15 - 16 | 1794 | 55 |
NM_178224.3 | NP_835742.1 | 15 - 16 | 1797 | 55 | |
GenBank mRNA | AK077669.1 | - | 15 - 16 | 1758 | 55 |
BC013472.1 | - | 15 - 16 | 1723 | 55 | |
BC026595.1 | - | 15 - 16 | 1730 | 55 |