Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- 4930453H23Rik
- Entrez Gene ID
-
100503481
- Gene Name
- RIKEN cDNA 4930453H23 gene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 21206580 - 21221672 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CB274217.1 |
- |
2 - 3 |
437 |
|
Chr. X - 21218410 - 21221564 [-] on Build GRCm38 |
67 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
synovial sarcoma, X member A, breakpoint 1 |
Ssxa1 |
Gm1532, Ssxa |
-96.3kb
|
Anti-sense |
Chr. - 21115816 - 21122088 on Build GRCm38 |
Downstream |
predicted gene 6938 |
Gm6938 |
EG629024 |
92.8kb
|
Sense |
Chr. - 21312210 - 21334727 on Build GRCm38 |
View Full Product Details