Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grik1
Entrez Gene ID
14805
Gene Name
glutamate receptor, ionotropic, kainate 1
Gene Aliases
A830007B11Rik, D16Ium24, D16Ium24e, GluK1, GluK5, Glur-5, Glur5, Glurbeta1
Chromosome Location
Chr.16: 87895897 - 88290602 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK044420.1 - 15 - 16 3073 103
AK044706.1 - 2 - 3 1006 103
BC031822.1 - 14 - 15 2540 103
BC049275.1 - 14 - 15 2540 103

View Full Product Details