Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdk9
Entrez Gene ID
107951
Gene Name
cyclin-dependent kinase 9 (CDC2-related kinase)
Gene Aliases
PITALRE
Chromosome Location
Chr.2: 32705782 - 32712784 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130860.3 NP_570930.1 6 - 7 884 150
GenBank mRNA AF327431.1 - 6 - 7 756 150
AK089276.1 - 6 - 7 723 150
AK142397.1 - 4 - 5 919 150
AK143217.1 - 6 - 7 885 150
AK144607.1 - 6 - 7 865 150
AK155733.1 - 4 - 5 1252 150
AK157340.1 - 6 - 7 856 150
BC003901.1 - 6 - 7 829 150

View Full Product Details