The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013372.2 | NP_001013390.1 | 1 - 1 | 350 | 82 |
GenBank mRNA | AK012518.1 | - | 1 - 1 | 169 | 82 |
AK049549.1 | - | 1 - 1 | 173 | 82 | |
AY753183.1 | - | 1 - 1 | 350 | 82 | |
BC119399.1 | - | 1 - 1 | 327 | 82 | |
BC119401.1 | - | 1 - 1 | 327 | 82 |