The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009749.2 | NP_033879.1 | 3 - 3 | 391 | 66 |
GenBank mRNA | AF097439.1 | - | 3 - 3 | 265 | 66 |
AK132005.1 | - | 3 - 3 | 278 | 66 | |
BC027529.1 | - | 3 - 3 | 265 | 66 |