Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Socs4
Entrez Gene ID
67296
Gene Name
suppressor of cytokine signaling 4
Gene Aliases
3110032M18Rik, A730004F22Rik, AI427843, AU015727, AU015859, Socs7
Chromosome Location
Chr.14: 47277143 - 47291591 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK080408.1 - - 1153 85


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN677544.1 - 1 Chr. 14 - 47294333 - 47294994 [-] on Build GRCm38 85
GenBankmRNA CV675449.1 - 1 791 Chr. 14 - 47294004 - 47294827 [+] on Build GRCm38 85
GenBankmRNA BM894612.1 - 1 254 Chr. 14 - 47294511 - 47294987 [+] on Build GRCm38 85
GenBankmRNA DT903349.1 - 1 604 Chr. 14 - 47294160 - 47294953 [+] on Build GRCm38 85
GenBankmRNA BX525705.1 - 1 133 Chr. 14 - 47294632 - 47295163 [+] on Build GRCm38 85
GenBankmRNA BI145262.1 - 1 417 Chr. 14 - 47294392 - 47295023 [+] on Build GRCm38 85
GenBankmRNA AI553361.1 - 1 132 Chr. 14 - 47294633 - 47295111 [+] on Build GRCm38 85
GenBankmRNA AA529335.1 - 1 132 Chr. 14 - 47294633 - 47295178 [+] on Build GRCm38 85


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Downstream mitogen-activated protein kinase 1 interacting protein 1-like Mapk1ip1l 9130202B12, C130032J12Rik 2.9kb Sense Chr. - 47297880 - 47323091 on Build GRCm38

View Full Product Details