Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Snhg14
- Entrez Gene ID
-
52480
- Gene Name
- small nucleolar RNA host gene 14
- Gene Aliases
-
AU018661, D7Ertd715e, Lncat
- Chromosome Location
-
Chr.7: 59969577 - 59974431 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK080843.1 |
- |
- |
1816 |
102 |
BC070450.1 |
- |
- |
313 |
102 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CD803437.1 |
- |
2 - 3 |
313 |
|
Chr. 7 - 59971900 - 59974257 [-] on Build GRCm38 |
102 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small nucleolar RNA, C/D box 116-like 1 |
Snord116l1 |
Pwcr1l1 |
-78.6kb
|
Sense |
Chr. - 59894622 - 59894711 on Build GRCm38 |
Downstream |
small nucleolar RNA, C/D box 64 |
Snord64 |
MBII-13 |
4.9kb
|
Sense |
Chr. - 59978811 - 59978856 on Build GRCm38 |
View Full Product Details