The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022004.6 | NP_071287.1 | 1 - 2 | 304 | 99 |
GenBank mRNA | AK003888.1 | - | 1 - 2 | 70 | 99 |
BC051127.1 | - | 1 - 2 | 78 | 99 | |
BY081454.1 | - | 1 - 2 | 305 | 99 | |
BY704145.1 | - | 1 - 2 | 64 | 99 |