Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klra23
Entrez Gene ID
79410
Gene Name
killer cell lectin-like receptor subfamily A, member 23
Gene Aliases
Ly-49W1, Ly-49W2, Ly49W
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024470.1 NP_077790.1 5 - 6 687 71
GenBank mRNA AF074459.1 - 5 - 6 687 71
AF074463.1 - 5 - 6 678 71
AF283250.1 - 4 - 5 570 71
AF283251.1 - 4 - 5 561 71

Gene Symbol
Klra13-ps
Entrez Gene ID
16631
Gene Name
killer cell lectin-like receptor subfamily A, member 13, pseudogene
Gene Aliases
Klra13, Ly49M
Chromosome Location
Chr.6: 130291161 - 130306432 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033451.1 4 - 5 71
GenBank mRNA AF074460.1 - 4 - 5 71
AF283252.1 - - 71
BC099861.1 - - 71

Gene Symbol
Klra7
Entrez Gene ID
16638
Gene Name
killer cell lectin-like receptor, subfamily A, member 7
Gene Aliases
LGL-1, Ly-49G.1, Ly-49G.2, Ly-49G.3, Ly49G.4, Ly49g, Ly49g1, Ly49g2
Chromosome Location
Chr.6: 130218605 - 130233322 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110323.1 NP_001103793.1 6 - 7 730 71
NM_014194.5 NP_055009.5 4 - 5 631 71
GenBank mRNA AF074457.1 - 4 - 5 568 71
AF283248.1 - 5 - 6 607 71
AF283249.1 - 4 - 5 568 71
AF283253.1 - 3 - 4 478 71
AF288375.1 - 5 - 6 600 71
AF288376.1 - 4 - 5 590 71
AF307945.1 - 5 - 6 607 71
AF307946.1 - 4 - 5 568 71
AF419249.1 - - 850 71
AK143240.1 - 4 - 5 590 71
BC099859.1 - 4 - 5 567 71
BC099860.1 - 4 - 5 567 71
BC099862.1 - 4 - 5 567 71
BC130251.1 - 4 - 5 616 71
BC140984.1 - 4 - 5 614 71
EU429321.1 - 4 - 5 561 71
GU434662.1 - 4 - 5 561 71
GU434663.1 - 4 - 5 561 71
S78689.1 - 4 - 5 590 71
U10093.1 - 6 - 7 730 71
U10094.1 - 4 - 5 631 71
U12890.1 - 4 - 5 582 71

View Full Product Details