Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olfr69
Entrez Gene ID
18370
Gene Name
olfactory receptor 69
Gene Aliases
3'[b]3, 3'beta3, MOR22-2
Chromosome Location
Chr.7: 103767277 - 103771594 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013621.3 NP_038649.2 2 - 3 173 88

View Full Product Details