Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUNDC2
Entrez Gene ID
101104126
Gene Name
FUN14 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.X: 86386418 - 86410044 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012107034.3 XP_011962424.2 4 - 5 514 88

Gene Symbol
LOC114118711
Entrez Gene ID
114118711
Gene Name
FUN14 domain-containing protein 2
Gene Aliases
-
Chromosome Location
Chr.16: 2037289 - 2038266 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979967.1 XP_027835768.1 1 - 1 487 88

View Full Product Details