Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS5
Entrez Gene ID
101112174
Gene Name
integrator complex subunit 5
Gene Aliases
-
Chromosome Location
Chr.21: 42846781 - 42851408 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019604.4 XP_004019653.2 1 - 2 125 79

View Full Product Details