Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISOC2
Entrez Gene ID
101120012
Gene Name
isochorismatase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.14: 66743117 - 66749474 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978778.1 XP_027834579.1 2 - 3 233 77

View Full Product Details