Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKAP12
Entrez Gene ID
101105509
Gene Name
A-kinase anchoring protein 12
Gene Aliases
-
Chromosome Location
Chr.8: 82426610 - 82536527 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972558.1 XP_027828359.1 3 - 4 551 68

View Full Product Details