Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL3RA
Entrez Gene ID
101113991
Gene Name
interleukin 3 receptor subunit alpha
Gene Aliases
-
Chromosome Location
Chr.X: 763904 - 783265 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963322.1 XP_027819123.1 7 - 8 884 98
XM_027963323.1 XP_027819124.1 7 - 8 1142 98

View Full Product Details