Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHISA7
Entrez Gene ID
101120268
Gene Name
shisa family member 7
Gene Aliases
-
Chromosome Location
Chr.14: 66756562 - 66767283 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978779.1 XP_027834580.1 3 - 4 989 69

View Full Product Details