Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD27
Entrez Gene ID
101114260
Gene Name
CD27 molecule
Gene Aliases
-
Chromosome Location
Chr.3: 224695208 - 224700443 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006941.4 XP_004006990.1 5 - 6 1093 59

View Full Product Details