Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCBP2
Entrez Gene ID
101102358
Gene Name
nuclear cap binding protein subunit 2
Gene Aliases
-
Chromosome Location
Chr.1: 209444604 - 209451335 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003016.4 XP_004003065.1 2 - 3 337 113

View Full Product Details