Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNMA1
Entrez Gene ID
100008745
Gene Name
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases
BKCA, KCNMA, RBSLO1
Chromosome Location
Chr.18: 10127405 - 10672476 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082070.1 NP_001075539.1 24 - 25 3070 69
GenBank mRNA AB009312.1 - 23 - 24 2884 69
AF201702.1 - 24 - 25 3070 69
AF321818.1 - 23 - 24 3148 69

View Full Product Details