Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARG2
Entrez Gene ID
100008967
Gene Name
arginase 2
Gene Aliases
-
Chromosome Location
Chr.20: 4603546 - 4639377 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082181.1 NP_001075650.1 7 - 8 859 124
GenBank mRNA AY864853.1 - 7 - 8 859 124

View Full Product Details