Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FUNDC2
Entrez Gene ID
100328769
Gene Name
FUN14 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.Unknown: 697134 - 707065 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171385.1 NP_001164856.1 - 137 87

View Full Product Details