Gene Transcripts

Gene Symbol
Sptbn2
Entrez Gene ID
29211
Gene Name
spectrin, beta, non-erythrocytic 2
Gene Aliases
Spnb3
Chromosome Location
Chr. 1: 219964429 - 220006802 on Build Rnor_6.0
UniGene ID
Rn.20389
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019167.1 NP_062040.1 35 7244
XM_008760092.1 XP_008758314.1 34 7476
XM_008760093.1 XP_008758315.1 34 7476
XM_008760094.1 XP_008758316.1 35 6973
GenBank mRNA AB001347.1 - 35 7244
AB008551.1 - 34 6853
AF225960.1 - 34 6853

View Full Product Details