Gene Transcripts

Gene Symbol
Tdpoz1
Entrez Gene ID
365854
Gene Name
TD and POZ domain containing 1
Gene Aliases
Chromosome Location
Chr. 2: 194618508 - 194619601 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_345239.7 XP_345240.7 1 634

View Full Product Details