Gene Transcripts

Gene Symbol
Vom1r1
Entrez Gene ID
494234
Gene Name
vomeronasal 1 receptor 1
Gene Aliases
V1re5
Chromosome Location
Chr. Un|NW_007906788: 119045 - 119941 on Build Rnor_6.0
UniGene ID
Rn.129464
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009514.1 NP_001009514.1 607
GenBank mRNA AY510320.1 - 607

View Full Product Details