Gene Transcripts

Gene Symbol
Lrtm2
Entrez Gene ID
680883
Gene Name
leucine-rich repeats and transmembrane domains 2
Gene Aliases
Chromosome Location
Chr. 4: 151375568 - 151391153 on Build Rnor_6.0
UniGene ID
Rn.34565
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109430.1 NP_001102900.1 5 1421
XM_006237208.2 XP_006237270.1 6 2074

View Full Product Details