Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOC
Entrez Gene ID
704406
Gene Name
myocilin
Gene Aliases
-
Chromosome Location
Chr.1: 28959250 - 28975470 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001099905.4 XP_001099905.2 - 814 80
GenBank mRNA JV489117.1 - - 627 80

View Full Product Details