Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIEZO1
Entrez Gene ID
711216
Gene Name
piezo type mechanosensitive ion channel component 1
Gene Aliases
FAM38A
Chromosome Location
Chr.20: 75733325 - 75800475 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028841099.1 XP_028696932.1 - 4469 150

View Full Product Details