Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GALNT8
Entrez Gene ID
721912
Gene Name
polypeptide N-acetylgalactosaminyltransferase 8
Gene Aliases
-
Chromosome Location
Chr.11: 4815455 - 4879579 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015150879.2 XP_015006365.2 - 1371 150

View Full Product Details