Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC20
Entrez Gene ID
710594
Gene Name
leucine rich repeat containing 20
Gene Aliases
-
Chromosome Location
Chr.9: 67408482 - 67495495 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001108339.4 XP_001108339.1 - 104 104
XM_015147359.2 XP_015002845.1 - 104 104
GenBank mRNA JU321506.1 - - 80 104
JU476235.1 - - 74 104

View Full Product Details