Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN12
Entrez Gene ID
704058
Gene Name
claudin 12
Gene Aliases
EGK_13954, claudin-12
Chromosome Location
Chr.3: 115661034 - 115674496 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194860.2 NP_001181789.1 - 207 100
XM_015134056.2 XP_014989542.1 - 302 100
GenBank mRNA JU319945.1 - - 224 100
JV045133.1 - - 116 100

View Full Product Details