Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arg2
Entrez Gene ID
29215
Gene Name
arginase 2
Gene Aliases
-
Chromosome Location
Chr.6: 102311097 - 102338406 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019168.1 NP_062041.1 3 - 4 465 123
GenBank mRNA U90887.1 - 3 - 4 465 123

View Full Product Details