The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019279.1 | NP_062152.1 | 1 - 2 | 136 | 83 |
GenBank mRNA | AF181561.1 | - | 1 - 2 | 136 | 83 |
FQ214158.1 | - | 1 - 2 | 224 | 83 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008773095.1 | XP_008771317.1 | - | 1048 | 83 |