Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Egfl7
Entrez Gene ID
245963
Gene Name
EGF-like-domain, multiple 7
Gene Aliases
-
Chromosome Location
Chr.3: 4034762 - 4044280 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139104.1 NP_620804.1 2 - 3 287 97
XM_006233668.2 XP_006233730.1 2 - 3 467 97
GenBank mRNA AF223678.1 - 2 - 3 287 97
BC078725.1 - 2 - 3 308 97

View Full Product Details