Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chrnb1
Entrez Gene ID
24261
Gene Name
cholinergic receptor, nicotinic, beta 1 (muscle)
Gene Aliases
Acrb, RNACRB1
Chromosome Location
Chr.10: 56390671 - 56403226 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012528.1 NP_036660.1 6 - 7 677 69
XM_008767770.1 XP_008765992.1 6 - 7 724 69
GenBank mRNA X74833.1 - 6 - 7 677 69

View Full Product Details