The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006252530.2 | XP_006252592.1 | 6 - 7 | 513 | 74 |
XM_006252531.2 | XP_006252593.1 | 5 - 6 | 510 | 71 |