Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Doxl1
Entrez Gene ID
312316
Gene Name
diamine oxidase-like protein 1
Gene Aliases
-
Chromosome Location
Chr.4: 78563625 - 78567241 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199233.1 NP_954703.1 3 - 4 1980 85

View Full Product Details