Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhoh
Entrez Gene ID
305341
Gene Name
ras homolog family member H
Gene Aliases
-
Chromosome Location
Chr.14: 43961756 - 43992587 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013430.1 NP_001013448.1 1 - 2 246 70
GenBank mRNA BC091269.1 - 1 - 2 246 70
FQ225173.1 - 1 - 2 257 70
FQ228192.1 - 1 - 2 336 70
FQ232983.1 - 1 - 2 257 70

View Full Product Details