Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdk9
Entrez Gene ID
362110
Gene Name
cyclin-dependent kinase 9
Gene Aliases
-
Chromosome Location
Chr.3: 11742269 - 11747117 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007743.1 NP_001007744.1 3 - 4 309 97
GenBank mRNA BC082037.1 - 3 - 4 309 97

View Full Product Details