Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smpd2
Entrez Gene ID
83537
Gene Name
sphingomyelin phosphodiesterase 2, neutral membrane
Gene Aliases
nSMase
Chromosome Location
Chr.20: 46212070 - 46215236 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031360.2 NP_112650.1 4 - 5 570 66
XM_008773007.1 XP_008771229.1 5 - 6 507 66
GenBank mRNA AB047002.1 - 4 - 5 309 66
BC091139.1 - 4 - 5 570 66

View Full Product Details