Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lipa
Entrez Gene ID
25055
Gene Name
lipase A, lysosomal acid, cholesterol esterase
Gene Aliases
Chole, Chole2, LAL, Lip1
Chromosome Location
Chr.1: 252816536 - 252870302 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012732.3 NP_036864.3 3 - 4 511 87
XM_006231275.2 XP_006231337.1 3 - 4 478 87
XM_008760321.1 XP_008758543.1 2 - 3 374 87
GenBank mRNA BC072532.1 - 3 - 4 511 87

View Full Product Details