Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jsrp1
Entrez Gene ID
690423
Gene Name
junctional sarcoplasmic reticulum protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 11769950 - 11774373 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109591.1 NP_001103061.1 3 - 4 213 69

View Full Product Details