The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105777.2 | NP_001099247.2 | 12 - 13 | 1463 | 65 |
GenBank mRNA | BC161929.1 | - | 12 - 13 | 1449 | 65 |
CF110623.1 | - | 8 - 9 | 942 | 65 |