Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif19
Entrez Gene ID
303659
Gene Name
kinesin family member 19
Gene Aliases
Kif19a, RGD1559936
Chromosome Location
Chr.10: 103308392 - 103334466 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271202.1 NP_001258131.1 8 - 9 925 62

View Full Product Details