Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a12
Entrez Gene ID
365398
Gene Name
solute carrier family 22 (organic anion/urate transporter), member 12
Gene Aliases
Rst, Slc22al2
Chromosome Location
Chr.1: 221910787 - 221919276 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034943.1 NP_001030115.1 1 - 2 662 65
XM_008760161.1 XP_008758383.1 2 - 3 1829 65
XM_008760162.1 XP_008758384.1 2 - 3 1677 65
GenBank mRNA BC103638.1 - 1 - 2 662 65

View Full Product Details