Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rasa4
Entrez Gene ID
288589
Gene Name
RAS p21 protein activator 4
Gene Aliases
Capri, RGD1565457
Chromosome Location
Chr.12: 23596989 - 23624388 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002727953.3 XP_002727999.1 9 - 10 1026 139

View Full Product Details