The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168529.1 | NP_001162001.1 | 20 - 21 | 2349 | 66 |
XM_006234956.2 | XP_006235018.1 | 23 - 24 | 2747 | 66 | |
XM_008762158.1 | XP_008760380.1 | 20 - 21 | 3091 | 66 |