The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134990.1 | NP_001128462.1 | 4 - 5 | 699 | 156 |
XM_006252059.2 | XP_006252121.1 | 5 - 6 | 1552 | 156 | |
XM_006252061.2 | XP_006252123.1 | 4 - 5 | 1380 | 156 | |
GenBank mRNA | BC168660.1 | - | 4 - 5 | 699 | 156 |
FQ230419.1 | - | 5 - 6 | 749 | 156 |