Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc56
Entrez Gene ID
365389
Gene Name
leucine rich repeat containing 56
Gene Aliases
RGD1311654
Chromosome Location
Chr.1: 214181751 - 214197184 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024902.1 NP_001020073.1 3 - 4 341 70
XM_006230564.2 XP_006230626.1 3 - 4 827 70
XM_006230565.2 XP_006230627.1 3 - 4 937 70
XM_006230566.2 XP_006230628.1 3 - 4 1110 70
XM_008760005.1 XP_008758227.1 3 - 4 1109 70
GenBank mRNA BC097444.1 - 3 - 4 341 70

View Full Product Details