Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp6v1c1
Entrez Gene ID
299971
Gene Name
ATPase, H+ transporting, lysosomal V1 subunit C1
Gene Aliases
-
Chromosome Location
Chr.7: 77678864 - 77716658 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011992.1 NP_001011992.1 9 - 10 827 113
XM_006241602.2 XP_006241664.1 8 - 9 832 113
GenBank mRNA BC089961.1 - 9 - 10 827 113

View Full Product Details